Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SSSCA1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584661
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584661 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584661 Supplier Invitrogen™ Supplier No. PA584661
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VMACTQTALL QKLTWASAEL GSSTSLETSI QLCGLIRACA EALRSLQQLQ H Highest antigen sequence indentity to the following orthologs: Rat - 86%, Mouse - 86%.

Might play a role in mitosis. Antigenic molecule. Could be a centromere-associated protein. May induce anti-centromere antibodies.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SSSCA1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNRD2
Gene Accession No. O60232
Gene Alias 1500016H19Rik; Autoantigen p27; autoantigen p27 homolog; C184l; centromeric autoantigen (27kD); p27; Protein C184L; Protein ZNRD2; Sjoegren syndrome/scleroderma autoantigen 1; Sjoegren syndrome/scleroderma autoantigen 1 homolog; Sjogren syndrome/scleroderma autoantigen 1; Sjogren syndrome/scleroderma autoantigen 1 homolog; Sjogren's syndrome/scleroderma autoantigen 1; Sjogren's syndrome/scleroderma autoantigen 1 homolog; Sjogren's syndrome/scleroderma autoantigen 1 homolog (human); SSSCA1; Zinc ribbon domain-containing protein 2; Znrd2
Gene Symbols ZNRD2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SSSCA1. Recombinant protein control fragment (Product #RP-108500).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10534
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.