Learn More
Invitrogen™ SSSCA1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA587664
Description
Immunogen sequence: MALNGAEVDD FSWEPPTEAE TKVLQARRER QDRISRLMGD YLLRGYRMLG ETCADCGTIL LQDKQRKIYC VACQELDSDV DKDNPALNAQ AALSQAREHQ LASASELPLG SRPAPQPPVP RPEHCEGAAA GLKAAQGPPA PAVPPNTDVM ACTQTALLQK LTWASAELGS STSLETSIQL CGLIRACAEA LRSLQQLQH; Positive Samples: Mouse liver.
Might play a role in mitosis. Antigenic molecule. Could be a centromere-associated protein. May induce anti-centromere antibodies.
Specifications
SSSCA1 | |
Polyclonal | |
Unconjugated | |
ZNRD2 | |
1500016H19Rik; Autoantigen p27; autoantigen p27 homolog; C184l; centromeric autoantigen (27kD); p27; Protein C184L; Protein ZNRD2; Sjoegren syndrome/scleroderma autoantigen 1; Sjoegren syndrome/scleroderma autoantigen 1 homolog; Sjogren syndrome/scleroderma autoantigen 1; Sjogren syndrome/scleroderma autoantigen 1 homolog; Sjogren's syndrome/scleroderma autoantigen 1; Sjogren's syndrome/scleroderma autoantigen 1 homolog; Sjogren's syndrome/scleroderma autoantigen 1 homolog (human); SSSCA1; Zinc ribbon domain-containing protein 2; Znrd2 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
56390 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.28 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P56873 | |
ZNRD2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-199 of human SSSCA1 (NP_0063871). | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.