Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SSTR1 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA3108

Catalog No. PA3108


Only null left
Edge
Add to Cart

Description

Description

PA3-108 detects the Somatostatin Receptor 1 in human samples. PA3-108 has been used successfully in Western blot, immunocytochemistry, and immunohistochemistry procedures. In Western blot analysis of SSTR1 transfected HEK-293 cells this antibody detects a ~52-63 kDa protein representing SSTR1. The PA3-108 immunogen is a synthetic peptide corresponding to residues C(380) R N G T C T S R I T T L(391) of human SSTR1.

Somatostatin acts to regulate numerous physiological processes by binding to and activating specific receptors in target tissues. Activation of these receptors by somatostatin, which is secreted by nerve and endocrine cells, regulates the secretion of insulin, glucagon and growth hormone, neuronal excitability in both the brain and the peripheral nervous system, and cell growth. Somatostatin receptors have been implicated in numerous diseases ranging from Alzheimer's to cancers of the gastrointestinal tract, breast, prostate, and pituitary.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SSTR1
Polyclonal
Unconjugated
SSTR1
Gpcrrna; RNGPCRRNA; Smstr1; Smstr-1; somatostatin receptor; somatostatin receptor 1; Somatostatin receptor subtype 1; somatostatin receptor type 1; SRIF-2; SS1R; SS-1-R; SS1-R; sst1; Sst1 receptor; Sstr1
Rabbit
100 μL
Primary
Human
Antibody
IgG
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Conc. Not Determined
Whole Serum with 0.05% sodium azide
P30872
SSTR1
Synthetic peptide corresponding to residues C(380) R N G T C T S R I T T L(391) of human SSTR1.
RUO
6751
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.