Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SSTR1/Somatostatin Receptor 1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981316
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1316 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1316 Supplier Bioss Supplier No. BS1137R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Somatostatin acts to regulate numerous physiological processes by binding to and activating specific receptors in target tissues. Activation of these receptors by somatostatin, which is secreted by nerve and endocrine cells, regulates the secretion of insulin, glucagon and growth hormone, neuronal excitability in both the brain and the peripheral nervous system, and cell growth. Somatostatin receptors have been implicated in numerous diseases ranging from Alzheimer′s to cancers of the gastrointestinal tract, breast, prostate, and pituitary.

Specifications

Antigen SSTR1/Somatostatin Receptor 1
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene SSTR1
Gene Accession No. P30872
Gene Alias Gpcrrna; RNGPCRRNA; Smstr1; Smstr-1; somatostatin receptor 1; Somatostatin receptor subtype 1; Somatostatin receptor type 1; SRIF-2; SS1R; SS-1-R; SS1-R; sst1; Sst1 receptor; SSTR1
Gene Symbols SSTR1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human SSTR1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6751
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.