Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SSTR2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA120630
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA120630 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA120630 Supplier Invitrogen™ Supplier No. PA120630
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Suggested positive controls are pancreas and islet of Langerhans.

SSTR2 (somatostatin receptor 2) is a member of the superfamily of receptors having seven transmembrane segments and is expressed in highest levels in the cerebrum and kidney. Somatostatin acts at many sites to inhibit the release of many hormones and other secretory proteins. The receptor is coupled via pertussis toxin sensitive G proteins to inhibit adenylyl cyclase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SSTR2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene SSTR2
Gene Accession No. P30680, P30874, P30875, P34993, P34994, Q49LX6
Gene Alias Smstr2; Smstr-2; somatostatin receptor; somatostatin receptor 2; somatostatin receptor 2a; somatostatin receptor subtype 2; somatostatin receptor type 2; somatotropin release-inhibiting factor receptor; SRIF-1; SS2R; SS-2-R; SS2-R; SSR2; sst2; Sstr2; SSTR-2; SSTR2A
Gene Symbols SSTR2
Host Species Rabbit
Immunogen Synthetic 17 amino acid peptide from 2nd extracellular domain of human SSTR2.
Purification Method Antigen affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100052744, 100356395, 100753171, 20606, 282083, 403472, 497059, 54305, 6752
Target Species Human, Mouse, Rat, Canine, Rabbit, Monkey, Bovine, Hamster, Pig, Horse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.