Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SSTR2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120328
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120328 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120328 Supplier Invitrogen™ Supplier No. PA5120328
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: mouse kidney, mouse liver. The target is usually found in the following locations: Cell membrane, Cytoplasm, Multi-pass membrane protein. Immunogen sequence: LPFYIFNVSS VSMAISPTPA LKGMFDFVVV LTYANSCANP ILYAFLSDNF KKSFQNVLCL VKVSGTDDGE RSDSKQDKSR LNETTETQRT LLNGDLQTSI.

SSTR2 (somatostatin receptor 2) is a member of the superfamily of receptors having seven transmembrane segments and is expressed in highest levels in the cerebrum and kidney. Somatostatin acts at many sites to inhibit the release of many hormones and other secretory proteins. The receptor is coupled via pertussis toxin sensitive G proteins to inhibit adenylyl cyclase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SSTR2
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.17 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene SSTR2
Gene Accession No. P30680, P30874, P30875
Gene Alias Smstr2; Smstr-2; somatostatin receptor; somatostatin receptor 2; somatostatin receptor 2a; somatostatin receptor subtype 2; somatostatin receptor type 2; somatotropin release-inhibiting factor receptor; SRIF-1; SS2R; SS-2-R; SS2-R; SSR2; sst2; Sstr2; SSTR-2; SSTR2A
Gene Symbols SSTR2
Host Species Rabbit
Immunogen Synthetic peptide.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20606, 54305, 6752
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.