Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SSTR2 Recombinant Superclonal™ Antibody (11HCLC)

Catalog No. PI711091
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI711091 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI711091 Supplier Invitrogen™ Supplier No. 711091
Add to Cart
Add to Cart

Rabbit Recombinant Superclonal Antibody

This antibody is predicted to react with Monkey. Recombinant rabbit Superclonal™ antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds - the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody - recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets - a recombinant rabbit Superclonal™ antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production. Note: Formerly called 'Recombinant polyclonal antibody', this product is now rebranded as 'Recombinant Superclonal™ antibody'. The physical product and the performance remain unchanged.

SSTR2 (somatostatin receptor 2) is a member of the superfamily of receptors having seven transmembrane segments and is expressed in highest levels in the cerebrum and kidney. Somatostatin acts at many sites to inhibit the release of many hormones and other secretory proteins. The receptor is coupled via pertussis toxin sensitive G proteins to inhibit adenylyl cyclase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SSTR2
Applications Western Blot, Immunocytochemistry
Classification Recombinant Superclonal
Clone 11HCLC
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2-7.4
Gene SSTR2
Gene Accession No. P30874
Gene Alias Smstr2; Smstr-2; somatostatin receptor; somatostatin receptor 2; somatostatin receptor 2a; somatostatin receptor subtype 2; somatostatin receptor type 2; somatotropin release-inhibiting factor receptor; SRIF-1; SS2R; SS-2-R; SS2-R; SSR2; sst2; Sstr2; SSTR-2; SSTR2A
Gene Symbols SSTR2
Host Species Rabbit
Immunogen Peptide corresponding to Human SSTR2 (aa 355-369).
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6752
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Do you offer SSTR2 antibodies for western blot detection?

We offer 5 SSTR2 polyclonal antibodies that target human and have been verified for western blot detection (https://www.thermofisher.com/antibody/primary/target/sstr2/filter/proteintarget/SSTR2/application/Western%20Blot/species/Human). Their tested dilutions for western blot detection are listed below.

Catalog Number and related western blot dilution information:
PA3-109    1:250
711091     1-2 µg/mL
PA3-16736   1:100-1:1000
PA5-85431   1:500-1:3000
BS-1138R   Assay Dependent

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.