Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SSTR2/Somatostatin Receptor 2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981326
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1326 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1326 Supplier Bioss Supplier No. BS1138R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SSTR2 (somatostatin receptor 2) is a member of the superfamily of receptors having seven transmembrane segments and is expressed in highest levels in the cerebrum and kidney. Somatostatin acts at many sites to inhibit the release of many hormones and other secretory proteins. The receptor is coupled via pertussis toxin sensitive G proteins to inhibit adenylyl cyclase.

Specifications

Antigen SSTR2/Somatostatin Receptor 2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene SSTR2
Gene Accession No. P30874
Gene Alias Smstr2; Smstr-2; somatostatin receptor 2; somatostatin receptor subtype 2; Somatostatin receptor type 2; somatotropin release-inhibiting factor receptor; SRIF-1; SS2R; SS-2-R; SS2-R; SSR2; Sst2; SSTR2; SSTR-2; SSTR2A
Gene Symbols SSTR2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human SSTR2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6752
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.