Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SSTR5 Mouse anti-Human, Mouse, Rat, Clone: 2B10D8, Proteintech

Mouse Monoclonal Antibody
Supplier: Proteintech Group Inc 667721IG150UL

Description
Somatostatin is a multifunctional protein affecting exocrine and endocrine protein secretion, neurotransmission, and neuronal modulation and regulating cell proliferation. All these functions of Somatostatin are executed through a class of GPCR protein family members called somatostatin receptor. Till date, 6 classes of somatostatin receptors have been discovered out of which SSTR5, 364A.A protein, is a receptor subtype expressed predomitly in the pituitary and hypothalamus region of brain. Studies revealed that SSTR5 contains seven transmembrane regions and undergoes homodimerization and also heterodimerizes with SSTR1. Although the specific functions of SSTR5 are still elusive, the studies indicate that it complements SSTR2 regulation of ACTH secretion. Polymorphisms in SSTR5 gene have been observed in patients with acromegaly.Specifications
SSTR5 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O08858, P30938, P35346 | |
SSTR5 | |
SSTR5 Fusion Protein Ag18615 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |
Western Blot | |
2B10D8 | |
Unconjugated | |
SSTR5 | |
somatostatin receptor 5, Somatostatin receptor type 5, SS 5 R, SS5 R, SS5R, SSTR5 | |
Mouse | |
Protein G | |
RUO | |
20609, 25354, 6755 | |
-20°C | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction