Learn More
Invitrogen™ SSTR5 Recombinant Rabbit Monoclonal Antibody (24GB2600)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA557729

Description
SSTR5 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Somatostatin is a multifunctional protein affecting exocrine and endocrine protein secretion, neurotransmission, and neuronal modulation and regulating cell proliferation. All these functions of Somatostatin are executed through a class of GPCR protein family members called somatostatin receptor. Till date, 6 classes of somatostatin receptors have been discovered out of which SSTR5, 364A.A protein, is a receptor subtype expressed predominantly in the pituitary and hypothalamus region of brain. Studies revealed that SSTR5 contains seven transmembrane regions and undergoes homodimerization and also heterodimerizes with SSTR1. Although the specific functions of SSTR5 are still elusive, the studies indicate that it complements SSTR2 regulation of ACTH secretion. Polymorphisms in SSTR5 gene have been observed in patients with acromegaly.
Specifications
SSTR5 | |
Recombinant Monoclonal | |
0.6 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O08858, P30938, P35346 | |
Sstr5 | |
A synthesized peptide derived from human SSTR5 (aa 1-64). | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Flow Cytometry, Western Blot, Immunocytochemistry | |
24GB2600 | |
Unconjugated | |
Sstr5 | |
Smstr5; Somato; somatostatin receptor; somatostatin receptor 5; Somatostatin receptor subtype 5; somatostatin receptor type 5; SS5R; SS-5-R; SS5-R; sst5; Sstr5 | |
Rabbit | |
Affinity chromatography | |
RUO | |
20609, 25354, 6755 | |
-20°C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.