Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ssu72 Antibody (66D10), Alexa Fluor™ 647, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP260236AF647
This item is not returnable.
View return policy
Description
Ssu72 Monoclonal antibody specifically detects Ssu72 in Human samples. It is validated for Western Blot, ELISASpecifications
Ssu72 | |
Monoclonal | |
Alexa Fluor 647 | |
CTD phosphatase SSU72, EC 3.1.3.16, FLJ13048, HSPC182, PNAS-120, RNA polymerase II subunit A C-terminal domain phosphatase SSU72, SSU72 RNA polymerase II CTD phosphatase homolog (S. cerevisiae), Ssu72 RNA polymerase II CTD phosphatase homolog (yeast) | |
Recombinant human SSU72 (1-194aa) purified from E. coli | |
0.1 mL | |
DNA replication Transcription Translation and Splicing, Proteases & Other Enzymes | |
29101 | |
Store at 4°C in the dark. | |
IgG2b κ |
Western Blot, ELISA | |
66D10 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction