Learn More
Invitrogen™ SSU72 Monoclonal Antibody (OTI2E2)

Description
SSU72 Monoclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | SSU72 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI2E2 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | Ssu72 |
| Gene Accession No. | Q9NP77 |
| Gene Alias | 1190002E22Rik; 1500011L16Rik; 2610101M12Rik; CTD phosphatase SSU72; FLJ13048; HSPC182; PNAS-120; RGD1307854; RNA polymerase II subunit A C-terminal domain phosphatase SSU72; RP5-832C2.2; SSU72; SSU72 homolog, RNA polymerase II CTD phosphatase; SSU72 RNA polymerase II CTD phosphatase homolog (S. cerevisiae); Ssu72 RNA polymerase II CTD phosphatase homolog (yeast) |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.