Learn More
SSU72 Mouse anti-Human, Clone: OTI5A1, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF809190

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Human Ssu72 is a highly conserved homologue of yeast Ssu72, a CTD phosphatase and a component of the polyadenylation/ termination machinery. The human Ssu72 is located on Chromosome 1p36 and seems to represent a single gene. It was identified as a pRb binding factor and was also shown to interact with TFIIB and yeast PTA1.Specifications
SSU72 | |
Monoclonal | |
Unconjugated | |
SSU72 | |
HSPC182, PNAS-120 | |
Mouse | |
Affinity Chromatography | |
RUO | |
29101 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI5A1 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9NP77 | |
SSU72 | |
Full length human recombit protein of human SSU72 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.