Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SSU72 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578483
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578483 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578483 Supplier Invitrogen™ Supplier No. PA578483
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: 293T, A431, HeLa, HepG2 Predicted Reactivity: Mouse (99%), Zebrafish (90%), Xenopus laevis (93%), Dog (98%), Chicken (98%), Rhesus Monkey (100%), Bovine (99%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

SSU72 is a highly conserved homologue of yeast Ssu72, a CTD phosphatase and a component of the polyadenylation/ termination machinery. The human Ssu72 is located on Chromosome 1p36 and seems to represent a single gene. It was identified as a pRb binding factor and was also shown to interact with TFIIB and yeast PTA1. The ABs recognize a single band of approximately 30 Kda on immunoblots of various human cell lines but do not work well on mouse tissues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SSU72
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.27 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene Ssu72
Gene Accession No. Q4KLK9, Q9CY97, Q9NP77
Gene Alias 1190002E22Rik; 1500011L16Rik; 2610101M12Rik; CTD phosphatase SSU72; FLJ13048; HSPC182; PNAS-120; RGD1307854; RNA polymerase II subunit A C-terminal domain phosphatase SSU72; RP5-832C2.2; SSU72; SSU72 homolog, RNA polymerase II CTD phosphatase; SSU72 RNA polymerase II CTD phosphatase homolog (S. cerevisiae); Ssu72 RNA polymerase II CTD phosphatase homolog (yeast)
Gene Symbols Ssu72
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human SSU72. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29101, 298681, 68991
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.