Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SSX1 Antibody (OTI1E10), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274362H
Description
SSX1 Monoclonal specifically detects SSX1 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
SSX1 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
SSX1 | |
Full length recombinant protein of human SSX1 (NP_005626) produced in E.coli. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI1E10 | |
Western Blot, Immunohistochemistry | |
Cancer/testis antigen 5.1, CT5.1cancer/testis antigen family 5, member 1, MGC150425, protein SSX1, sarcoma, synovial, X-chromosome-related 1, SSRC, synovial sarcoma, X breakpoint 1MGC5162 | |
Mouse | |
Affinity Purified | |
Stem Cell Markers | |
6756.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction