Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SSX2 Antibody (OTI4D10), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274368F
Description
SSX2 Monoclonal specifically detects SSX2 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
SSX2 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
SSX2 | |
Full-length protein expressed in 293T cell transfected with human SSX2 expression vector. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI4D10 | |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
Cancer/testis antigen 5.2, cancer/testis antigen family 5, member 2a, CT5.2a, HD21, HOM-MEL-40, MGC119055, MGC15364, MGC3884, protein SSX2, sarcoma, synovial, X-chromosome-related 2, SSX, SSX2A, SSX2B, synovial sarcoma, X breakpoint 2B, synovial sarcoma, X breakpoint 2CT5.2, Tumor antigen HOM-MEL-40 | |
Mouse | |
Affinity Purified | |
Cancer | |
6757.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction