Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SSX2IP Antibody (OTI5C3), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274369F
Description
SSX2IP Monoclonal specifically detects SSX2IP in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.Specifications
SSX2IP | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
SSX2IP | |
Recombinant protein fragment corresponding to amino acids 1-272 of human SSX2IP (NP_054740) produced in E.coli. | |
0.1 mL | |
117178.0 | |
Store at 4°C in the dark. |
Immunohistochemistry | |
OTI5C3 | |
Immunohistochemistry | |
ADIP, afadin- and alpha-actinin-binding protein, Afadin DIL domain-interacting protein, FLJ10848, KIAA0923, MGC75026, SSX2-interacting protein, synovial sarcoma, X breakpoint 2 interacting protein | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction