Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SSX2IP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531495
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531495 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531495 Supplier Invitrogen™ Supplier No. PA531495
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: Neuro2A, GL261, C8D30. Predicted reactivity: Mouse (95%), Rat (93%), Chicken (88%), Rhesus Monkey (100%), Chimpanzee (100%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a protein that binds the cancer-testis antigen Synovial Sarcoma X breakpoint 2 protein. The encoded protein may regulate the activity of Synovial Sarcoma X breakpoint 2 protein in malignant cells. Alternate splicing results in multiple transcript variants. A pseudogene of this gene is found on chromosome 3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SSX2IP
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene SSX2IP
Gene Accession No. Q8VC66, Q9Y2D8
Gene Alias ADIP; Afadin- and alpha-actinin-binding protein; afadin- and alpha-actinin-binding protein ADIP; Afadin- and alpha-actinin-binding protein-like protein; afadin DIL domain-interacting protein; AU014939; AU042321; FLJ10848; KIAA0923; LOW QUALITY PROTEIN: afadin- and alpha-actinin-binding protein; MGC75026; SSX family member 2 interacting protein; SSX2-interacting protein; Ssx2ip; synovial sarcoma, X breakpoint 2 interacting protein
Gene Symbols SSX2IP
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human SSX2IP. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117178, 99167
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.