Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SSX3 Monoclonal Antibody (4A11)
GREENER_CHOICE

Catalog No. PIMA524431
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524431 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524431 Supplier Invitrogen™ Supplier No. MA524431
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: MNGDDTFARR PTVGAQIPEK IQKAFDDIAK YFSKEEWEKM KVSEKIVYVY MKRKYEAMTK LGFKAILPSF MRNKRVTDFQ GNDFDNDPNR GNQVQRPQMT.

Could act as a modulator of transcription.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SSX3
Applications ELISA, Western Blot
Classification Monoclonal
Clone 4A11
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene SSX3
Gene Accession No. Q99909
Gene Alias cancer/testis antigen 5.3; CT5.3; CT53; Kruppel-associated box containing gene product SSX3; protein SSX3; SSX family member 3; SSX3; synovial sarcoma, X breakpoint 3
Gene Symbols SSX3
Host Species Mouse
Immunogen SSX3 (NP_066294, 1 a.a. ~ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10214
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.