Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SSX3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569016
Description
This target displays homology in the following species: Cow: 80%; Dog: 85%; Guinea Pig: 83%; Horse: 83%; Human: 100%; Mouse: 86%; Pig: 100%; Rabbit: 85%; Rat: 100%.
Could act as a modulator of transcription.
Specifications
SSX3 | |
Polyclonal | |
Unconjugated | |
SSX3 | |
cancer/testis antigen 5.3; CT5.3; CT53; Kruppel-associated box containing gene product SSX3; protein SSX3; SSX family member 3; SSX3; synovial sarcoma, X breakpoint 3 | |
Rabbit | |
Affinity chromatography | |
RUO | |
10214 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q99909 | |
SSX3 | |
synthetic peptide directed towards the N terminal region of human SSX3. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction