Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SSX4B Monoclonal Antibody (OTI2E10), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA812589

Description
SSX4B belongs to the family of highly homologous synovial sarcoma X (SSX) breakpoint proteins. These proteins may function as transcriptional repressors. They are also capable of eliciting spontaneously humoral and cellular immune responses in cancer patients, and are potentially useful targets in cancer vaccine-based immunotherapy. SSX1, SSX2 and SSX4 genes have been involved in the t(X;18) translocation characteristically found in all synovial sarcomas. This translocation results in the fusion of the synovial sarcoma translocation gene on chromosome 18 to one of the SSX genes on chromosome X. Chromosome Xp11 contains a segmental duplication resulting in two identical copies of synovial sarcoma, X breakpoint 4, SSX4 and SSX4B, in tail-to-tail orientation. This gene, SSX4B, represents the more centromeric copy. Two transcript variants encoding distinct isoforms have been identified for this gene.Specifications
SSX4B | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
CT5.4 | |
Mouse | |
Affinity Chromatography | |
RUO | |
548313 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
OTI2E10 | |
Unconjugated | |
SSX4B | |
SSX4B | |
Full length human recombit protein of human SSX4B produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction