Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SSX5 Antibody (OTI3G6), PE/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274372PECY7
Description
SSX5 Monoclonal specifically detects SSX5 in Human samples. It is validated for Flow Cytometry.Specifications
SSX5 | |
Monoclonal | |
PE-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
SSX5 | |
Full length recombinant protein of human SSX5 (NP_066295) produced in HEK293T cells. | |
0.1 mL | |
6758.0 | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
OTI3G6 | |
Flow Cytometry | |
MGC9494, protein SSX5, synovial sarcoma, X breakpoint 5 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction