Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SSX5 Monoclonal Antibody (OTI1D6), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA504221S


Description
SSX5 Monoclonal antibody specifically detects SSX5 in Canine, Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
SSX5 | |
Monoclonal | |
0.78 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q7TP48, Q9D7N9, Q9HDC9 | |
Mouse | |
Affinity chromatography | |
RUO | |
366227, 57136, 608089, 71881 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG2b |
Western Blot | |
OTI1D6 | |
Unconjugated | |
APMAP | |
protein SSX5; SSX family member 5; SSX5; synovial sarcoma, X breakpoint 5 | |
Full length human recombinant protein of human C20orf3 produced in HEK293T cell. | |
30 μL | |
Primary | |
Canine, Human, Mouse, Rat | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction