Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ ST 034307
GREENER_CHOICE

Catalog No. 627110
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
62-711-0 10 mg
62-715-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 62-711-0 Supplier Tocris Bioscience™ Supplier No. 6271/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Adenylyl cyclase 1 (AC1) inhibitor; analgesic and active in vivo

ST 034307 is an adenylyl cyclase 1 (AC1) inhibitor (IC50 = 2.3 μM). Exhibits some selectivity for AC1 over other AC isoforms, with no significant activity against AC8. Inhibits Ca2+/calmodulin-stimulated cAMP accumulation in hippocampal homogenates. Enhances μ opioid receptor (MOR)-mediated inhibition of AC1 in cellular assays and inhibits development and maintenance of MOR-mediated sensitization of AC1. ST 034307 relieves pain without causing tolerance in mouse models of pain. Active in vivo. Analgesic.

Chemical Identifiers

CAS 133406-29-8
Molecular Formula C10H4Cl4O2

Specifications

Quantity 10 mg
Solubility Soluble to 100mM in DMSO and to 20mM in ethanol
Formula Weight 297.95
Physical Form Solid
Purity >98%
Molecular Weight (g/mol) 297.94
Chemical Name or Material 6-Chloro-2-(trichloromethyl)-4H-1-benzopyran-4-one
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.