Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ ST 045849
GREENER_CHOICESDP

Catalog No. 677510 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
67-751-0 10 mg
67-755-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 67-751-0 Supplier Tocris Bioscience™ Supplier No. 6775/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

O-GlcNAc transferase (OGT) inhibitor

ST 045849 is an O-GlcNAc transferase (OGT) inhibitor (IC50 = 53 μM). Reduces proliferation and viability of prostate cancer cells in vitro. Depletes intracellular alanine and decreases glucose consumption by cancer cells. Decreases markers of ER stress and apoptosis in pregnant diabetic mice and reduces neural tube defects in embryos from diabetic mice.

Specifications

Chemical Name or Material 2-[(4-Chlorophenyl)imino]tetrahydro-4-oxo-3-(2-tricyclo[3.3.1.13,7]dec-1-ylethyl)-2H-1,3-thiazine-6-carboxylic acid
CAS 442665-87-4
Quantity 10 mg
Molecular Formula C23H27ClN2O3S
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.