Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ST14 Monoclonal Antibody (416802)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA524154
Description
This antibody detects human Matriptase/ST14 Catalytic Domain in direct ELISAs and Western blots. Reconstitute at 0.5 mg/mL in sterile PBS.
SOX2 is an intronless gene encoding a member of the SRY-related HMG-box (SOX) family of transcription factors involved in the regulation of embryonic development and in the determination of cell fate. The product of the SOX2 gene is required for stem-cell maintenance in the central nervous system, and also regulates gene expression in the stomach. The SOX2 gene lies within an intron of another gene called SOX2 overlapping transcript (SOX2OT). Further, SOX2 protein may act as a transcriptional activator after forming a protein complex with other proteins. Mutations in the SOX2 gene have been associated with bilateral anophthalmia, a severe form of structural eye malformation, optic nerve hypoplasia and syndromic microphthalmia.
Specifications
ST14 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q9Y5Y6 | |
ST14 | |
E. coli-derived recombinant human Matriptase/ST14 Catalytic Domain Val615-Val855. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Western Blot | |
416802 | |
Unconjugated | |
ST14 | |
ARCI11; Epithin; HAI; LOW QUALITY PROTEIN: suppressor of tumorigenicity 14 protein; matriptase; matriptase/MT-SP1; mCAP3; membrane-type serine protease; membrane-type serine protease 1; MTSP1; MT-SP1; MTSP-1; Prostamin; protease, serine, 14 (epithin); PRSS14; Serine protease 14; serine protease TADG-15; SNC19; ST14; ST14 transmembrane serine protease matriptase; suppression of tumorigenicity 14; suppression of tumorigenicity 14 (colon carcinoma); suppression of tumorigenicity 14 (colon carcinoma, matriptase, epithin); suppressor of tumorigenicity 14 protein; Suppressor of tumorigenicity 14 protein homolog; suppressor of tumorigenicity protein 14; TADG15; TADG-15; Tmprss14; tumor associated differentially expressed gene 15 protein; tumor-associated differentially-expressed gene 15 protein | |
Mouse | |
Protein A/G | |
RUO | |
6768 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction