Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ST14 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547554
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547554 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547554 Supplier Invitrogen™ Supplier No. PA547554
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

This antibody detects human Matriptase/ST14 Catalytic Domain in direct ELISAs and Western blots. In direct ELISAs, approximately 60% cross-reactivity with recombinant mouse Matriptase/Epithin is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

SOX2 is an intronless gene encoding a member of the SRY-related HMG-box (SOX) family of transcription factors involved in the regulation of embryonic development and in the determination of cell fate. The product of the SOX2 gene is required for stem-cell maintenance in the central nervous system, and also regulates gene expression in the stomach. The SOX2 gene lies within an intron of another gene called SOX2 overlapping transcript (SOX2OT). Further, SOX2 protein may act as a transcriptional activator after forming a protein complex with other proteins. Mutations in the SOX2 gene have been associated with bilateral anophthalmia, a severe form of structural eye malformation, optic nerve hypoplasia and syndromic microphthalmia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ST14
Applications Flow Cytometry, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene ST14
Gene Accession No. Q9Y5Y6
Gene Alias ARCI11; Epithin; HAI; LOW QUALITY PROTEIN: suppressor of tumorigenicity 14 protein; matriptase; matriptase/MT-SP1; mCAP3; membrane-type serine protease; membrane-type serine protease 1; MTSP1; MT-SP1; MTSP-1; Prostamin; protease, serine, 14 (epithin); PRSS14; Serine protease 14; serine protease TADG-15; SNC19; ST14; ST14 transmembrane serine protease matriptase; suppression of tumorigenicity 14; suppression of tumorigenicity 14 (colon carcinoma); suppression of tumorigenicity 14 (colon carcinoma, matriptase, epithin); suppressor of tumorigenicity 14 protein; Suppressor of tumorigenicity 14 protein homolog; suppressor of tumorigenicity protein 14; TADG15; TADG-15; Tmprss14; tumor associated differentially expressed gene 15 protein; tumor-associated differentially-expressed gene 15 protein
Gene Symbols ST14
Host Species Sheep
Immunogen E. coli-derived recombinant human Matriptase/ST14 Catalytic Domain Val615-Val855.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6768
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.