Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ST18 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116320
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116320 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116320 Supplier Invitrogen™ Supplier No. PA5116320
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ST18 Polyclonal Antibody for Western Blot

Repressor that binds to DNA sequences containing a bipartite element consisting of a direct repeat of the sequence 5'-AAAGTTT-3' separated by 2-9 nucleotides. Represses basal transcription activity from target promoters. Inhibits colony formation in cultured breast cancer cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ST18
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene ST18
Gene Accession No. Q80TY4
Gene Alias AV348974; C2-HC type zinc finger protein r-MyT3; Kiaa0535; mKIAA0535; Myt3; Neural zinc finger factor 3; neural zinc finger protein NZF-3; neural zinc finger transcription factor 3; Nzf3; NZF-3; r-MyT3; St18; ST18 C2H2C-type zinc finger transcription factor; ST18, C2H2C-type zinc finger; suppression of tumorigenicity 18; suppression of tumorigenicity 18 (breast carcinoma) (zinc finger protein); suppression of tumorigenicity 18 protein; suppression of tumorigenicity 18, zinc finger; suppression of tumorigenicity protein 18; ZC2H2C3; ZC2HC10; zinc finger protein 387; ZNF387
Gene Symbols ST18
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 195-222 amino acids from the N-terminal region of mouse St18.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 240690
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.