Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ST2/IL-33R Antibody (2154B), DyLight 405, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals FAB5232E
Description
ST2/IL-33R Monoclonal antibody specifically detects ST2/IL-33R in Human samples. It is validated for Flow Cytometry, ELISASpecifications
ST2/IL-33R | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, ELISA | |
2154B | |
Flow Cytometry, ELISA | |
DER4ST2growth stimulation-expressed, FIT-1, IL33R, interleukin 1 receptor-like 1, interleukin 1 receptor-related protein, interleukin-1 receptor-like 1, MGC32623, Protein ST2, ST2L, ST2V, T1homolog of mouse growth stimulation-expressed | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human ST-2/ IL-33R, Lys19-Phe328, Accession # Q01638 | |
0.1 mL | |
Angiogenesis | |
9173 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction