Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ST3 beta-Gal alpha-2,3-Sialyltransferase 2/ST3GAL2 Antibody, Janelia Fluor™ 646, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298478JF646
Description
ST3 beta-Gal alpha-2,3-Sialyltransferase 2/ST3GAL2 Polyclonal antibody specifically detects ST3 beta-Gal alpha-2,3-Sialyltransferase 2/ST3GAL2 in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
ST3 beta-Gal alpha-2,3-Sialyltransferase 2/ST3GAL2 | |
Polyclonal | |
Western Blot, Immunoprecipitation | |
Alpha 2,3-ST 2, Beta-galactoside alpha-2,3-sialyltransferase 2, beta-galactoside alpha-2,3-sialytransferase, CMP-N-acetylneuraminate-beta-galactosamide-alpha-2,3-sialyltransferase 2, EC 2.4.99.4, EC 2.4.99.7, Gal-beta-1,3-GalNAc-alpha-2,3-sialyltransferase, Gal-NAc6S, Sialyltransferase 4B, sialyltransferase 4B (beta-galactosidase alpha-2,3-sialytransferase), SIAT4B, SIAT4-B, ST3 beta-galactoside alpha-2,3-sialyltransferase 2, ST3GalA.2ST3Gal II, ST3GALII | |
Produced in rabbits immunized with E. coli-derived Human ST3 beta-Gal alpha-2,3-Sialyltransferase 2/ST3GAL2 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, Immunoprecipitation | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
6483 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction