Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ST3GAL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542928
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542928 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542928 Supplier Thermo Scientific Supplier No. PA542928
Add to Cart
Add to Cart

ST3GAL2 Polyclonal Antibody

ST3GAL2 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen ST3GAL2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ST3GAL2
Gene Accession No. Q16842
Gene Alias 3-sialytransferase; AI429591; alpha 2,3-sialyltransferase ST3Gal II; alpha 2,3-ST 2; alpha-2,3-sialyltransferase; AW822065; beta-galactoside alpha-2,3-sialyltransferase 2; CMP-N-acetylneuraminate-beta-galactosamide-alpha-2,3-sialyltransferase 2; Gal-beta-1,3-GalNAc-alpha-2,3-sialyltransferase; gal-NAc6S; si:ch211-223o1.7; sialyltransferase 4B; sialyltransferase 4B (beta-galactosidase alpha-2,3-sialytransferase); sialyltransferase 4B (beta-galactoside alpha-2,3-sialyltransferase); sialyltransferase 5; sialyltransferase ST3Gal-II; Siat4b; SIAT4-B; siat5; ST3 beta-galactoside alpha-2,3-sialyltransferase 2; ST3Gal II; st3Gal II.2; St3gal2; st3gal2-r1; ST3GALA.2; ST3GalII
Gene Symbols ST3GAL2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human ST3GAL2 (aa 301-350).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6483
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.