Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ST3GAL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112275
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112275 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112275 Supplier Invitrogen™ Supplier No. PA5112275
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ST3GAL2 Polyclonal Antibody for Western Blot, IP

ST3GAL2 is a type II membrane protein that catalyzes the transfer of sialic acid from CMP-sialic acid to galactose-containing substrates. The protein is normally found in the Golgi but can be proteolytically processed to a soluble form. This protein, which is a member of glycosyltransferase family 29, can use the same acceptor substrates as does sialyltransferase 4A. The protein encoded by this gene is a type II membrane protein that catalyzes the transfer of sialic acid from CMP-sialic acid to galactose-containing substrates. The encoded protein is normally found in the Golgi but can be proteolytically processed to a soluble form. This protein, which is a member of glycosyltransferase family 29, can use the same acceptor substrates as does sialyltransferase 4A.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ST3GAL2
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.03% ProClin 300; pH 7
Gene ST3GAL2
Gene Accession No. Q16842
Gene Alias 3-sialytransferase; AI429591; alpha 2,3-sialyltransferase ST3Gal II; alpha 2,3-ST 2; alpha-2,3-sialyltransferase; AW822065; beta-galactoside alpha-2,3-sialyltransferase 2; CMP-N-acetylneuraminate-beta-galactosamide-alpha-2,3-sialyltransferase 2; Gal-beta-1,3-GalNAc-alpha-2,3-sialyltransferase; gal-NAc6S; si:ch211-223o1.7; sialyltransferase 4B; sialyltransferase 4B (beta-galactosidase alpha-2,3-sialytransferase); sialyltransferase 4B (beta-galactoside alpha-2,3-sialyltransferase); sialyltransferase 5; sialyltransferase ST3Gal-II; Siat4b; SIAT4-B; siat5; ST3 beta-galactoside alpha-2,3-sialyltransferase 2; ST3Gal II; st3Gal II.2; St3gal2; st3gal2-r1; ST3GALA.2; ST3GalII
Gene Symbols ST3GAL2
Host Species Rabbit
Immunogen E. coli-derived Human ST3GAL2 fragment.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6483
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.