Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ST3GAL3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
$382.00 - $610.00
Specifications
Antigen | ST3GAL3 |
---|---|
Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
ST3GAL3 Polyclonal specifically detects ST3GAL3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
ST3GAL3 | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
6487 | |
This antibody was developed against a recombinant protein corresponding to the amino acids: KLHLLQWEEDSNSVVLSFDSAGQTLGSEYDRLGFLLNLDSKLPAELATKYANFSEGACKPGYASALMTAIF | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot 0.4 ug/ml, Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
Polyclonal | |
Rabbit | |
Human | |
alpha 2,3-sialyltransferase III, Alpha 2,3-ST 3, alpha-2,3-sialyltransferase II, Beta-galactoside alpha-2,3-sialyltransferase 3, CMP-N-acetylneuraminate-beta-1,4-galactoside alpha-2,3-sialyltransferase, EC 2.4.99.6, Gal beta-1,3(4) GlcNAc alpha-2,3 sialyltransferase, Gal beta-1,3(4)GlcNAc alpha-2,3 sialyltransferase, N-acetyllactosaminide alpha-2,3-sialyltransferase, Sialyltransferase 6, SIAT6sialyltransferase 6 (N-acetyllacosaminide alpha 2,3-sialyltransferase), ST3 beta-galactoside alpha-2,3-sialyltransferase 3, ST3Gal III, ST3GALII, ST3GalIII, ST3N | |
ST3GAL3 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title