Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ST3GAL3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA542912
Description
Peptide sequence: GFGYDMSTPN APLHYYETVR MAAIKESWTH NIQREKEFLR KLVKARVITD Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 92%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.
The protein encoded by this gene is a type II membrane protein that catalyzes the transfer of sialic acid from CMP-sialic acid to galactose-containing substrates. The encoded protein is normally found in the Golgi apparatus but can be proteolytically processed to a soluble form. This protein is a member of glycosyltransferase family 29. Multiple transcript variants encoding several different isoforms have been found for this gene.
Specifications
ST3GAL3 | |
Polyclonal | |
Unconjugated | |
ST3GAL3 | |
alpha 2,3-sialyltransferase III; alpha 2,3-ST 3; alpha-2,3-sialyltransferase II; beta-galactoside alpha-2,3-sialyltransferase 3; CMP-N-acetylneuraminate-beta-1,4-galactoside alpha-2,3-sialyltransferase; CMP-N-acetylneuraminate-beta-1,4-galactoside alpha-2,3-sialyltransferase soluble form; CMP-N-acetylneuraminate-beta-1,4-galactoside alpha-2,3-sialyltransferase; LOW QUALITY PROTEIN: CMP-N-acetylneuraminate-beta-1,4-galactoside alpha-2,3-sialyltransferase; EIEE15; gal beta-1,3(4) GlcNAc alpha-2,3 sialyltransferase; Gal beta-1,3(4)GlcNAc alpha-2,3 sialyltransferase; mental retardation, non-syndromic, autosomal recessive, 12; MRT12; N-acetyllacosaminide alpha 2,3-sialyltransferase; N-acetyllactosaminide alpha-2,3-sialyltransferase; sialyltransferase (N-acetyllacosaminide alpha 23-sialyltransferase); sialyltransferase 3; sialyltransferase 6; sialyltransferase 6 (N-acetyllacosaminide alpha 2,3-sialyltransferase); sialyltransferase 6(N-acetyllacosaminide alpha 23-sialyltransferase); sialyltransferase ST3Gal-III; Siat3; Siat6; ST3 beta-galactoside alpha-2,3-sialyltransferase 3; ST3Gal III; St3gal3; ST3GALII; ST3GalIII; ST3N | |
Rabbit | |
Protein A | |
RUO | |
6487 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q11203 | |
ST3GAL3 | |
Synthetic peptide directed towards the C-terminal of human ST3GAL3 (aa 321-370). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction