Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ST3GAL3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562317
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562317 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562317 Supplier Invitrogen™ Supplier No. PA562317
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KLHLLQWEED SNSVVLSFDS AGQTLGSEYD RLGFLLNLDS KLPAELATKY ANFSEGACKP GYASALMTAI F Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 90%.

The protein encoded by this gene is a type II membrane protein that catalyzes the transfer of sialic acid from CMP-sialic acid to galactose-containing substrates. The encoded protein is normally found in the Golgi apparatus but can be proteolytically processed to a soluble form. This protein is a member of glycosyltransferase family 29. Multiple transcript variants encoding several different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ST3GAL3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ST3GAL3
Gene Accession No. Q11203
Gene Alias alpha 2,3-sialyltransferase III; alpha 2,3-ST 3; alpha-2,3-sialyltransferase II; beta-galactoside alpha-2,3-sialyltransferase 3; CMP-N-acetylneuraminate-beta-1,4-galactoside alpha-2,3-sialyltransferase; CMP-N-acetylneuraminate-beta-1,4-galactoside alpha-2,3-sialyltransferase soluble form; CMP-N-acetylneuraminate-beta-1,4-galactoside alpha-2,3-sialyltransferase; LOW QUALITY PROTEIN: CMP-N-acetylneuraminate-beta-1,4-galactoside alpha-2,3-sialyltransferase; EIEE15; gal beta-1,3(4) GlcNAc alpha-2,3 sialyltransferase; Gal beta-1,3(4)GlcNAc alpha-2,3 sialyltransferase; mental retardation, non-syndromic, autosomal recessive, 12; MRT12; N-acetyllacosaminide alpha 2,3-sialyltransferase; N-acetyllactosaminide alpha-2,3-sialyltransferase; sialyltransferase (N-acetyllacosaminide alpha 23-sialyltransferase); sialyltransferase 3; sialyltransferase 6; sialyltransferase 6 (N-acetyllacosaminide alpha 2,3-sialyltransferase); sialyltransferase 6(N-acetyllacosaminide alpha 23-sialyltransferase); sialyltransferase ST3Gal-III; Siat3; Siat6; ST3 beta-galactoside alpha-2,3-sialyltransferase 3; ST3Gal III; St3gal3; ST3GALII; ST3GalIII; ST3N
Gene Symbols ST3GAL3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ST3GAL3. Recombinant protein control fragment (Product #RP-101251).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6487
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.