Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ST3GAL4 Antibody, mFluor Violet 500 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297314MFV500
Description
ST3GAL4 Polyclonal antibody specifically detects ST3GAL4 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
ST3GAL4 | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
Alpha 2,3-sialyltransferase IV, Alpha 2,3-ST 4, Beta-galactoside alpha-2,3-sialyltransferase 4, CGS23FLJ46764, EC 2.4.99, EC 2.4.99.-, EC 2.4.99.9, FLJ11867, gal-beta-1,4-GalNAc-alpha-2,3-sialyltransferase, Gal-NAc6S, NANTA3alpha-3-N-acetylneuraminyltransferase, SAT3, SAT-3, Sialyltransferase 4C, sialyltransferase 4C (beta-galactosidase alpha-2,3-sialytransferase), sialyltransferase 4C (beta-galactoside alpha-2,3-sialytransferase), SIAT4-C, SIAT4CCMP-N-acetylneuraminate-beta-galactosamide-alpha-2,3-sialyltransferase 4, ST3 beta-galactoside alpha-2,3-sialyltransferase 4, ST3Gal IV, ST3GalA.2, ST3GalIV, ST-4, STZSIAT4 | |
Produced in rabbits immunized with E. coli-derived Human ST3GAL4 fragment. | |
0.1 mL | |
Amino Acids Drugs and other small molecules, Cancer, Endocrinology, Signal Transduction | |
6484 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction