Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ST3GAL4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562056
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562056 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562056 Supplier Invitrogen™ Supplier No. PA562056
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SREDRYIELF YFPIPEKKEP CLQGEAESKA SKLFGNYSRD QPIFLRLEDY FWVKTPSAYE Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 82%.

Synthesis of alpha-2, 3-linked sialic acid to Gal(beta-1, 3)GalNAc is mediated by at least 3 distinct beta-galactoside alpha-2, 3-sialyltransferases (EC 2.4.99.4), including ST3GAL4. In contrast, only a single gene encodes the beta-galactoside alpha-2, 6-sialyltransferase, ST6GAL1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ST3GAL4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene St3gal4
Gene Accession No. Q11206
Gene Alias alpha 2,3-sialyltransferase IV; Alpha 2,3-ST 4; alpha-2,3-sialyltransferase; alpha2,3-sialyltransferase; alpha-2,3-sialyltransferase ST3Gal IV; alpha-3-N-acetylneuraminyltransferase; beta-galactoside alpha-2,3-sialyltransferase 4; CGS23; CMP-N-acetylneuraminate-beta-galactosamide-alpha-2,3-sialyltransferase 4; FLJ11867; gal-beta-1,4-GalNAc-alpha-2,3-sialyltransferase; Gal-NAc6S; glycosyltransferase; NANTA3; SAT3; SAT-3; sialyltransferase 4C; sialyltransferase 4C (beta-galactosidase alpha-2,3-sialytransferase); sialyltransferase 4C (beta-galactoside alpha-2,3-sialytransferase); SIAT4; SIAT4C; SIAT4-C; ST3 beta-galactoside alpha-2,3-sialyltransferase 4; ST3Gal IV; ST3GAL4; ST3GalA.2; ST3GalIV; ST3GAL-IV; ST-4; STZ
Gene Symbols St3gal4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ST3GAL4. Recombinant protein control fragment (Product #RP-101187).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6484
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.