Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ST3GAL4 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA589294
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589294 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589294 Supplier Thermo Scientific Supplier No. PA589294

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

Rabbit Polyclonal Antibody

Immunogen sequence: REDSFYFPIP EKKEPCLQGE AESKASKLFG NYSRDQPIFL RLEDYFWVKT PSAYELPYGT KGSEDLLLRV LAITSSSIPK NIQSLRCRRC VVVGNGHRLR NSSLGDAINK YDVVIRLNNA PVAGYEGDVG SKTTMRLFYP ESAHFDPKVE NNPDTLLVLV AFKAMDFHWI ETILSDKKRV RKGFWKQPPL IWDVNPKQIR ILNPFFMEIA ADKLLSLPMQ QPRKIKQKPT TGLLAITLAL H; Positive Samples: Mouse ovary, Rat liver; Cellular Location: Golgi apparatus, Golgi stack membrane, Single-pass type II membrane protein

Synthesis of alpha-2, 3-linked sialic acid to Gal(beta-1, 3)GalNAc is mediated by at least 3 distinct beta-galactoside alpha-2, 3-sialyltransferases (EC 2.4.99.4), including ST3GAL4. In contrast, only a single gene encodes the beta-galactoside alpha-2, 6-sialyltransferase, ST6GAL1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ST3GAL4
Applications Western Blot
Classification Polyclonal
Concentration 0.69 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide, pH 7.3
Gene St3gal4
Gene Accession No. P61131, Q91Y74
Gene Alias alpha 2,3-sialyltransferase IV, Alpha 2,3-ST 4, alpha-2,3-sialyltransferase, alpha2,3-sialyltransferase, alpha-2,3-sialyltransferase ST3Gal IV, alpha-3-N-acetylneuraminyltransferase, beta-galactoside alpha-2,3-sialyltransferase 4, CGS23, CMP-N-acetylneuraminate-beta-galactosamide-alpha-2,3-sialyltransferase 4, FLJ11867, gal-beta-1,4-GalNAc-alpha-2,3-sialyltransferase, Gal-NAc6S, glycosyltransferase, NANTA3, SAT3, SAT-3, sialyltransferase 4C, sialyltransferase 4C (beta-galactosidase alpha-2,3-sialytransferase), sialyltransferase 4C (beta-galactoside alpha-2,3-sialytransferase), SIAT4, SIAT4C, SIAT4-C, ST3 beta-galactoside alpha-2,3-sialyltransferase 4, ST3Gal IV, ST3GAL4, ST3GalA.2, ST3GalIV, ST3GAL-IV, ST-4, STZ
Gene Symbols St3gal4
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 27-267 of human ST3GAL4 (NP_0062691)
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20443, 363040
Target Species Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.