Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ST3GAL5 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA525730

Catalog No. PIPA525730


Only null left
Add to Cart

Description

Description

ST3GAL5 Polyclonal Antibody for Western Blot, IHC (P), Flow

Ganglioside GM3 is known to participate in the induction of cell differentiation, modulation of cell proliferation, maintenance of fibroblast morphology, signal transduction, and integrin-mediated cell adhesion. The protein encoded by this gene is a type II membrane protein which catalyzes the formation of GM3 using lactosylceramide as the substrate. The encoded protein is a member of glycosyltransferase family 29 and may be localized to the Golgi apparatus. Mutation in this gene has been associated with Amish infantile epilepsy syndrome. Transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ST3GAL5
Polyclonal
Unconjugated
ST3GAL5
[a]2; 3S-T; alpha 2,3-sialyltransferase V; alpha2,3-sialyltransferase; CMP-NeuAc:lactosylceramide alpha-2,3-sialyltransferase; Ganglioside GM3 synthase; GM3 synthase; GM3S; GM3-specific sialytransferase; lactosylceramide alpha-2,3-sialyltransferase; lactosylceramide alpha-2,3-sialyltransferas-like protein; mST3Gal V; SATI; Sialyltransferase 9; sialyltransferase 9 (CMP-NeuAc:lactosylceramide alpha-2,3-sialyltransferase); sialyltransferase 9 (CMP-NeuAc:lactosylceramide alpha-2,3-sialyltransferase; GM3 synthase); Siat9; SIATGM3S; ST3 beta-galactoside alpha-2,3-sialyltransferase 5; ST3 beta-galactoside alpha-23-sialyltransferase 5; ST3Gal V; ST3GAL5; ST3GalV; ST3GAL-V; UNQ2510/PRO5998
Rabbit
Antigen affinity chromatography, Protein A
RUO
8869
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.4
Q9UNP4
ST3GAL5
KLH conjugated synthetic peptide between 389-418 amino acids of human ST3GAL5.
400 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.