Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ST3GAL5 Polyclonal Antibody
ST3GAL5 Polyclonal Antibody
Supplier: Thermo Scientific PA542867
Description
ST3GAL5 Polyclonal Antibody for Western Blot, IHC

Specifications
ST3GAL5 | |
Polyclonal | |
Unconjugated | |
ST3GAL5 | |
[a]2; 3S-T; alpha 2,3-sialyltransferase V; alpha2,3-sialyltransferase; CMP-NeuAc:lactosylceramide alpha-2,3-sialyltransferase; Ganglioside GM3 synthase; GM3 synthase; GM3S; GM3-specific sialytransferase; lactosylceramide alpha-2,3-sialyltransferase; lactosylceramide alpha-2,3-sialyltransferas-like protein; mST3Gal V; SATI; Sialyltransferase 9; sialyltransferase 9 (CMP-NeuAc:lactosylceramide alpha-2,3-sialyltransferase); sialyltransferase 9 (CMP-NeuAc:lactosylceramide alpha-2,3-sialyltransferase; GM3 synthase); Siat9; SIATGM3S; ST3 beta-galactoside alpha-2,3-sialyltransferase 5; ST3 beta-galactoside alpha-23-sialyltransferase 5; ST3Gal V; ST3GAL5; ST3GalV; ST3GAL-V; UNQ2510/PRO5998 | |
Rabbit | |
Protein A | |
RUO | |
8869 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9UNP4 | |
ST3GAL5 | |
Synthetic peptide directed towards the N-terminal of human ST3GAL5 (aa 151-200). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction