Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ST3GAL5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567026
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567026 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567026 Supplier Invitrogen™ Supplier No. PA567026
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NKTTIRMTYPE GAPLSDLEYY SNDLFVAVLF KSVDFNWLQA MVKKETLPFW VRLFFWKQVA EKIPLQPKHF RILNPVIIKE TAFDILQYSE PQSRFWG Highest antigen sequence identity to the following orthologs - mouse 93%, rat 94%.

Ganglioside GM3 is known to participate in the induction of cell differentiation, modulation of cell proliferation, maintenance of fibroblast morphology, signal transduction, and integrin-mediated cell adhesion. The protein encoded by this gene is a type II membrane protein which catalyzes the formation of GM3 using lactosylceramide as the substrate. The encoded protein is a member of glycosyltransferase family 29 and may be localized to the Golgi apparatus. Mutation in this gene has been associated with Amish infantile epilepsy syndrome. Transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ST3GAL5
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ST3GAL5
Gene Accession No. Q9UNP4
Gene Alias [a]2; 3S-T; alpha 2,3-sialyltransferase V; alpha2,3-sialyltransferase; CMP-NeuAc:lactosylceramide alpha-2,3-sialyltransferase; Ganglioside GM3 synthase; GM3 synthase; GM3S; GM3-specific sialytransferase; lactosylceramide alpha-2,3-sialyltransferase; lactosylceramide alpha-2,3-sialyltransferas-like protein; mST3Gal V; SATI; Sialyltransferase 9; sialyltransferase 9 (CMP-NeuAc:lactosylceramide alpha-2,3-sialyltransferase); sialyltransferase 9 (CMP-NeuAc:lactosylceramide alpha-2,3-sialyltransferase; GM3 synthase); Siat9; SIATGM3S; ST3 beta-galactoside alpha-2,3-sialyltransferase 5; ST3 beta-galactoside alpha-23-sialyltransferase 5; ST3Gal V; ST3GAL5; ST3GalV; ST3GAL-V; UNQ2510/PRO5998
Gene Symbols ST3GAL5
Host Species Rabbit
Immunogen Recombinant Human ST3GAL5. Recombinant protein control fragment (Product #RP-105461).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8869
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.