Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ST3GAL5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 146141AP150UL

Description
Ganglioside GM3 is known to participate in the induction of cell differentiation, modulation of cell proliferation, maintece of fibroblast morphology, signal transduction, and integrin-mediated cell adhesion. The protein encoded by this gene is a type II membrane protein which catalyzes the formation of GM3 using lactosylceramide as the substrate. The encoded protein is a member of glycosyltransferase family 29 and may be localized to the Golgi apparatus. Mutation in this gene has been associated with Amish infantile epilepsy syndrome. Transcript variants encoding different isoforms have been found for this gene.Specifications
ST3GAL5 | |
Polyclonal | |
Unconjugated | |
ST3GAL5 | |
Ganglioside GM3 synthase, Sialyltransferase 9, SIAT9, SIATGM3S, ST3Gal V, ST3GAL5, ST3GalV | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20454, 83505, 8869 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O88829, Q68G12, Q9UNP4 | |
ST3GAL5 | |
ST3GAL5 Fusion Protein Ag6207 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction