Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ST5 Polyclonal Antibody
ST5 Polyclonal Antibody
Supplier: Thermo Scientific PA561331
Description
ST5 Polyclonal Antibody for ICC/IF, IHC

Specifications
ST5 | |
Polyclonal | |
Unconjugated | |
DENND2B | |
2010004M01Rik; 2610305K15Rik; DENN domain-containing protein 2B; DENN/MADD domain containing 2B; DENND2B; HeLa tumor suppression 1; HTS1; p126; p70; St5; suppression of tumorigenicity 5; suppression of tumorigenicity 5 protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6764 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P78524 | |
DENND2B | |
Recombinant protein corresponding to Human ST5. Recombinant protein control fragment (Product #RP-97437). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction