Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ST6 Gal Sialyltransferase 1/ST6GAL1/CD75 Antibody (036), Alexa Fluor™ 350, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289923AF350
Description
ST6 Gal Sialyltransferase 1/ST6GAL1/CD75 Monoclonal specifically detects ST6 Gal Sialyltransferase 1/ST6GAL1/CD75 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
ST6 Gal Sialyltransferase 1/ST6GAL1/CD75 | |
Monoclonal | |
Alexa Fluor 350 | |
B-cell antigen CD75, beta-galactoside alpha-2,6-sialyltransferase 1, CMP-N-acetylneuraminate beta-galactosamide alpha-2,6-sialyltransferase, CMP-N-acetylneuraminate-beta-galactosamide-alpha-2,6-sialyltransferase 1, EC 2.4.99.1, MGC48859, Sialyltransferase 1, sialyltransferase 1 (beta-galactoside alpha-2,6-sialyltransferase), sialyltransferase 1 (beta-galactoside alpha-2,6-sialytransferase), SIAT1Alpha 2,6-ST 1, ST6 beta-galactosamide alpha-2,6-sialyltranferase 1, ST6Gal I, ST6GalI, ST6N | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human ST6 Gal Sialyltransferase 1/ST6GAL1/CD75 (P15907; Lys27-Cys406). | |
0.1 mL | |
Golgi Apparatus Markers | |
6480 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
36 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction