Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ST6GAL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106647
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106647 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106647 Supplier Invitrogen™ Supplier No. PA5106647
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ST6GAL1.

The protein encoded by this gene is a type II membrane protein that catalyzes the transfer of sialic acid from CMP-sialic acid to galactose-containing substrates. The encoded protein, which is normally found in the Golgi but which can be proteolytically processed to a soluble form, is involved in the generation of the cell-surface carbohydrate determinants and differentiation antigens HB-6, CDw75, and CD76. This protein is a member of glycosyltransferase family 29. Three transcript variants encoding two different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ST6GAL1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ST6GAL1
Gene Accession No. P13721, P15907, Q64685
Gene Alias Alpha 2,6-ST 1; AW742324; B-cell antigen CD75; beta galactoside alpha 2,6 sialyltransferase 1; beta-galactoside alpha-2,6-sialyltransferase 1; CMP-N-acetylneuraminate beta-galactosamide alpha-2,6-sialyltransferase; CMP-N-acetylneuraminate-beta-galactosamide-alpha-2,6-sialyltransferase 1; Sialyltransferase 1; sialyltransferase 1 (beta-galactoside alpha-2,6-sialyltransferase); Sialyltransferase 1 (beta-galactoside alpha-2,6-sialytransferase); Sialyltransferase 1 (beta-galactoside alpha-26-sialytransferase); SIAT1; ST6 beta-galactosamide alpha-2,6-sialyltranferase 1; ST6 beta-galactoside alpha-2,6-sialyltransferase 1; ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 1; St6gal; ST6Gal I; St6gal1; ST6GalI; St6Gal-I; ST6N
Gene Symbols ST6GAL1
Host Species Rabbit
Immunogen A synthesized peptide derived from human ST6GAL1(Accession P15907), corresponding to amino acid residues K177-T227.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20440, 25197, 6480
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.