Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ST6GAL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569666
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569666 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569666 Supplier Invitrogen™ Supplier No. PA569666
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 86%; Horse: 79%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.

Transfers sialic acid from the donor of substrate CMP-sialic acid to galactose containing acceptor substrates. Has alpha-2,6-sialyltransferase activity toward oligosaccharides that have the Gal-beta-1,4-GlcNAc sequence at the non-reducing end of their carbohydrate groups, but it has weak or no activities toward glycoproteins and glycolipids.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ST6GAL2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ST6GAL2
Gene Accession No. Q76K27
Gene Alias alpha 2,6 sialyltransferase; alpha 2,6-ST 2; alpha-2,6-sialyltransferase; alpha-2,6-sialyltransferase ST6Gal II; beta galactoside alpha 2,6 sialyltransferase 2; beta-galactosamide alpha-2,6-sialyltransferase II; beta-galactoside alpha-2,6-sialyltransferase 2; beta-galactoside alpha-2,6-sialyltransferase II; C230064G14Rik; CMP-N-acetylneuraminate-beta-galactosamide-alpha-2,6-sialyltransferase 2; hST6Gal II; Kiaa1877; LOW QUALITY PROTEIN: beta-galactoside alpha-2,6-sialyltransferase 2; mKIAA1877; sialyltransferase; sialyltransferase 2; sialyltransferase 2 (monosialoganglioside sialyltransferase); SIAT2; ST6 beta-galactosamide alpha-2,6-sialyltranferase 2; ST6 beta-galactosamide alpha-2,6-sialyltranferase 2a; ST6 beta-galactoside alpha-2,6-sialyltransferase 2; ST6Gal II; st6gal2; st6gal2a; ST6GalII; ST6Gal-II
Gene Symbols ST6GAL2
Host Species Rabbit
Immunogen synthetic peptide corresponding to a region of mouse ST6GAL2 (aa 286-335).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 240119
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.