Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ST6GAL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553169
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553169 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553169 Supplier Invitrogen™ Supplier No. PA553169
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PSSLSFLETR RLLPVQGKQR AIMGAAHEPS PPGGLDARQA LPRAHPAGSF HAGPGDLQKW AQSQDGFEHK EFFSSQVGRK SQSAFYPEDD DYFFAAGQPG WHSHTQGTLG FPSPGEPGPR EGAFPAAQ Highest antigen sequence identity to the following orthologs: Mouse - 59%, Rat - 60%.

Transfers sialic acid from the donor of substrate CMP-sialic acid to galactose containing acceptor substrates. Has alpha-2,6-sialyltransferase activity toward oligosaccharides that have the Gal-beta-1,4-GlcNAc sequence at the non-reducing end of their carbohydrate groups, but it has weak or no activities toward glycoproteins and glycolipids.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ST6GAL2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ST6GAL2
Gene Accession No. Q96JF0
Gene Alias alpha 2,6 sialyltransferase; alpha 2,6-ST 2; alpha-2,6-sialyltransferase; alpha-2,6-sialyltransferase ST6Gal II; beta galactoside alpha 2,6 sialyltransferase 2; beta-galactosamide alpha-2,6-sialyltransferase II; beta-galactoside alpha-2,6-sialyltransferase 2; beta-galactoside alpha-2,6-sialyltransferase II; C230064G14Rik; CMP-N-acetylneuraminate-beta-galactosamide-alpha-2,6-sialyltransferase 2; hST6Gal II; Kiaa1877; LOW QUALITY PROTEIN: beta-galactoside alpha-2,6-sialyltransferase 2; mKIAA1877; sialyltransferase; sialyltransferase 2; sialyltransferase 2 (monosialoganglioside sialyltransferase); SIAT2; ST6 beta-galactosamide alpha-2,6-sialyltranferase 2; ST6 beta-galactosamide alpha-2,6-sialyltranferase 2a; ST6 beta-galactoside alpha-2,6-sialyltransferase 2; ST6Gal II; st6gal2; st6gal2a; ST6GalII; ST6Gal-II
Gene Symbols ST6GAL2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ST6GAL2. Recombinant protein control fragment (Product #RP-90309).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84620
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.