Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ST6GAL2 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA572132
Description
Transfers sialic acid from the donor of substrate CMP-sialic acid to galactose containing acceptor substrates. Has alpha-2,6-sialyltransferase activity toward oligosaccharides that have the Gal-beta-1,4-GlcNAc sequence at the non-reducing end of their carbohydrate groups, but it has weak or no activities toward glycoproteins and glycolipids.
Specifications
ST6GAL2 | |
Polyclonal | |
Unconjugated | |
ST6GAL2 | |
Beta-galactoside alpha-2,6-sialyltransferase 2; Alpha 2,6-ST 2; CMP-N-acetylneuraminate-beta-galactosamide-alpha-2,6-sialyltransferase 2; ST6Gal II; ST6GalII; hST6Gal II; Sialyltransferase 2; ST6GAL2; KIAA1877; SIAT2 | |
Rabbit | |
Protein A | |
RUO | |
84620 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.3 mg/mL | |
PBS with 0.09% sodium azide | |
Q96JF0 | |
ST6GAL2 | |
KLH conjugated synthetic peptide between 483-509 amino acids from the C-terminal region of human ST6GAL2 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction