Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ST6GALNAC2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA544085
Description
Peptide sequence: VNTMKNSLVS YWNLGFTSVP QGQDLQYIFI PSDIRDYVML RSAILGVPVP Sequence homology: Cow: 91%; Dog: 85%; Horse: 100%; Human: 100%; Mouse: 92%; Pig: 100%; Rabbit: 100%; Rat: 100%.
ST6GALNAC2 belongs to a family of sialyltransferases that add sialic acids to the nonreducing ends of glycoconjugates. At the cell surface, these modifications have roles in cell-cell and cell-substrate interactions, bacterial adhesion, and protein targeting (Samyn-Petit et al., 2000 [PubMed 10742600]).
Specifications
ST6GALNAC2 | |
Polyclonal | |
Unconjugated | |
ST6GALNAC2 | |
(alpha-N-acetylneuraminyl-2,3-beta-galactosyl-1,3)-N-acetyl galactosaminide alpha-2,6-sialyltransferase; (alpha-N-acetylneuraminyl-2,3-beta-galactosyl-1,3)-N-acetyl galactosaminide B; alpha-N-acetylgalactosaminide alpha-2,6-sialyltransferase 2; alpha-N-acetylgalactosaminide alpha-2,6-sialyltransferase II; alpha-N-acetylgalactosaminide alpha-2,6-sialyltransferase II (); Gal-beta-1,3-GalNAc alpha-2,6-sialyltransferase; GalNAc alpha-2,6-sialyltransferase II; II; SAITL1; sialyltransferase 7; sialyltransferase 7 ((alpha-N-acetylneuraminyl 2,3-beta-galactosyl-1,3)-N-acetyl galactosaminde alpha-2,6-sialyltransferase) B; sialyltransferase 7 ((alpha-N-acetylneuraminyl-2,3-beta-galactosyl-1,3)-N-acetyl galactosaminide alpha-2,6-sialyltransferase) B; sialyltransferase 7 ((alpha-N-acetylneuraminyl-2,3-beta-galactosyl-1,3)-N-acetyl galactosaminide B; sialyltransferase 7B; sialyltransferase-like 1; Siat7; SIAT7B; SIAT7-B; SIATL1; ST6 (alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminide alpha-2,6-sialyltransferase 2; ST6 GalNAc alpha-2,6-sialyltransferase 2; ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 2; ST6GalNAc; ST6GalNAc II; St6galnac2; ST6GALNAC2, alpha-2,6-sialyltransferase 2; ST6GalNAcII; ST6GalNAII; SThM | |
Rabbit | |
Affinity chromatography | |
RUO | |
10610 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9UJ37 | |
ST6GALNAC2 | |
Synthetic peptide directed towards the middle region of human ST6GALNAC2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction