Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ST6GALNAC2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569976
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569976 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569976 Supplier Invitrogen™ Supplier No. PA569976
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Dog: 79%; Guinea Pig: 79%; Horse: 79%; Human: 100%; Mouse: 86%; Rabbit: 79%; Rat: 86%.

ST6GALNAC2 belongs to a family of sialyltransferases that add sialic acids to the nonreducing ends of glycoconjugates. At the cell surface, these modifications have roles in cell-cell and cell-substrate interactions, bacterial adhesion, and protein targeting (Samyn-Petit et al., 2000 [PubMed 10742600]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen ST6GALNAC2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ST6GALNAC2
Gene Accession No. 0
Gene Alias (alpha-N-acetylneuraminyl-2,3-beta-galactosyl-1,3)-N-acetyl galactosaminide alpha-2,6-sialyltransferase; (alpha-N-acetylneuraminyl-2,3-beta-galactosyl-1,3)-N-acetyl galactosaminide B; alpha-N-acetylgalactosaminide alpha-2,6-sialyltransferase 2; alpha-N-acetylgalactosaminide alpha-2,6-sialyltransferase II; alpha-N-acetylgalactosaminide alpha-2,6-sialyltransferase II (); Gal-beta-1,3-GalNAc alpha-2,6-sialyltransferase; GalNAc alpha-2,6-sialyltransferase II; II; SAITL1; sialyltransferase 7; sialyltransferase 7 ((alpha-N-acetylneuraminyl 2,3-beta-galactosyl-1,3)-N-acetyl galactosaminde alpha-2,6-sialyltransferase) B; sialyltransferase 7 ((alpha-N-acetylneuraminyl-2,3-beta-galactosyl-1,3)-N-acetyl galactosaminide alpha-2,6-sialyltransferase) B; sialyltransferase 7 ((alpha-N-acetylneuraminyl-2,3-beta-galactosyl-1,3)-N-acetyl galactosaminide B; sialyltransferase 7B; sialyltransferase-like 1; Siat7; SIAT7B; SIAT7-B; SIATL1; ST6 (alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminide alpha-2,6-sialyltransferase 2; ST6 GalNAc alpha-2,6-sialyltransferase 2; ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 2; ST6GalNAc; ST6GalNAc II; St6galnac2; ST6GALNAC2, alpha-2,6-sialyltransferase 2; ST6GalNAcII; ST6GalNAII; SThM
Gene Symbols ST6GALNAC2
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human ST6GALNAC2.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 303692
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.